accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOOOOH3CCH3OOSOOCH3

CAS RN 51064-65-4

IUPAC Name: (2,2-dimethyl-5-{[(4-methylbenzenesulfonyl)oxy]methyl}-1,3-dioxolan-4-yl)methyl 4-methylbenzene-1-sulfonate
Synonyms: (2,2-dimethyl-5-{[(4-methylbenzenesulfonyl)oxy]methyl}-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 470.55
Molecular Formula: C21H26O8S2
InChi Key: KPFDKWNWYAXRNJ-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1OC(C)(C)OC1COS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results