CAS RN 510758-28-8
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97+%
CAS: 510758-28-8 Molecular Formula: C30H30N10 Molecular Weight (g/mol): 530.64 MDL Number: MFCD09265124 InChI Key: WKGZJBVXZWCZQC-UHFFFAOYSA-N Synonym: tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta PubChem CID: 11203363 IUPAC Name: 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine SMILES: C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
CAS: 510758-28-8 Molecular Formula: C30H30N10 Molecular Weight (g/mol): 530.64 MDL Number: MFCD09265124 InChI Key: WKGZJBVXZWCZQC-UHFFFAOYSA-N Synonym: tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta PubChem CID: 11203363 IUPAC Name: 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine SMILES: C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Tocris Bioscience™ TBTA
Click chemistry auxiliary reagent; stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reaction
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
CAS: 510758-28-8 Molecular Formula: C30H30N10 Molecular Weight (g/mol): 530.64 MDL Number: MFCD09265124 InChI Key: WKGZJBVXZWCZQC-UHFFFAOYSA-N Synonym: tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta PubChem CID: 11203363 IUPAC Name: 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine SMILES: C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97.0+%, TCI America™
CAS: 510758-28-8 Molecular Formula: C30H30N10 Molecular Weight (g/mol): 530.64 MDL Number: MFCD09265124 InChI Key: WKGZJBVXZWCZQC-UHFFFAOYSA-N Synonym: tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine,1,2,4-trimethyl-5-benzenesulfonic acid,tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine,tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta PubChem CID: 11203363 IUPAC Name: tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine SMILES: C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
Sigma Aldrich Tris[(1-Benzyl-1H-1,2,3-Triazol-4-Yl)Methyl]Amine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.