Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TBTA
GREENER_CHOICESDP

Catalog No. 7862100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-621-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-621-00 Supplier R&D Systems™ Supplier No. 7862/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Click chemistry auxiliary reagent; stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reaction

TBTA is a click chemistry auxiliary reagent. It is a stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reactions.

Specifications

Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
CAS 510758-28-8
Quantity 100 mg
Product Line Tocris
Target General Laboratory Research Reagent
Molecular Formula C30H30N10
Purity 0.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.