accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOO

CAS RN 515-46-8

IUPAC Name: ethyl benzenesulfonate
Synonyms: ethyl benzenesulfonate
Molecular Weight (g/mol): 186.23
Molecular Formula: C8H10O3S
InChi Key: XDRMBCMMABGNMM-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results