CAS RN 516-72-3
IUPAC Name:
(1S,3aS,3bS,7S,9aR,9bS,11aS)-1-[(2S)-2-hydroxy-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms:
20 α-hydroxycholesterol 20R-hydroxycholesterol
Molecular Weight (g/mol):
402.66
Molecular Formula:
C27H46O2
InChi Key:
MCKLJFJEQRYRQT-APGJSSKUSA-N
SMILES:
CC(C)CCC[C@](C)(O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
1
–
1
of
1
results
1
R&D Systems™ 20(S)-Hydroxycholesterol
CAS: 516-72-3 Molecular Formula: C27H46O2