Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 20(S)-Hydroxycholesterol
GSA_VASDP

Catalog No. 447410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-741-0 Supplier R&D Systems™ Supplier No. 4474/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Allosteric activator of Hedgehog (Hh) signaling; induces Smo accumulation

Allosteric activator of the Hedgehog signaling pathway Smoothened (Smo) oncoprotein; binds at a site distinct from the canonical cyclopamine binding site. Activates Hedgehog (Hh) signaling (EC50 ∽ 3μM for induction of Hh reporter gene transcription in in NIH 3T3 cells). Induces Smo accumulation in primary cilia; also exhibits osteogenic activity and activates the liver X receptor (LXR).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 516-72-3
Molecular Formula C27H46O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 402.65
Purity >95%
Molecular Weight (g/mol) 402.663
Chemical Name or Material 20(S)-Hydroxycholesterol
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.