CAS RN 5162-03-8
CAS RN 5162-03-8
IUPAC Name:
(2-chlorophenyl)(phenyl)methanone
Synonyms:
(2-chlorophenyl)(phenyl)methanone
Molecular Weight (g/mol):
216.66
Molecular Formula:
C13H9ClO
InChi Key:
VMHYWKBKHMYRNF-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC=C1C(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
2-Chlorobenzophenone, 99+%
CAS: 5162-03-8 Molecular Formula: C13H9ClO Molecular Weight (g/mol): 216.66 MDL Number: MFCD00000558 InChI Key: VMHYWKBKHMYRNF-UHFFFAOYSA-N
2
2-Chlorobenzophenone 98.0+%, TCI America™

CAS: 5162-03-8 Molecular Formula: C13H9ClO Molecular Weight (g/mol): 216.66 MDL Number: MFCD00000558 InChI Key: VMHYWKBKHMYRNF-UHFFFAOYSA-N Synonym: 2-chlorobenzophenone,2-chlorophenyl phenyl methanone,methanone, 2-chlorophenyl phenyl,o-chlorobenzophenone,2-chlorophenyl phenylmethanone,benzophenone, 2-chloro,2-chlorophenyl-phenylmethanone,2-chlorobenyl phenyl ketone,2-chloro-phenyl-phenyl-methanone,benzophenone, 2-chloro-6ci,7ci,8ci PubChem CID: 78838 IUPAC Name: (2-chlorophenyl)-phenylmethanone SMILES: C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl