accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO(R)OH(S)(S)O(R)H3C(S)(S)OH(S)H3C

CAS RN 52-21-1

IUPAC Name: 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms: prednisolone acetate flo-pred omnipred predicort prenema meticortelone acetate pricortin supercortyl deltilen durapred
Molecular Weight (g/mol): 402.49
Molecular Formula: C23H30O6
InChi Key: LRJOMUJRLNCICJ-JZYPGELDSA-N
SMILES: CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Grade 
  • (1)
  • (5)
Percent Purity 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)