Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisolone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014436 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4436 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4436 Supplier MilliporeSigma Supelco Supplier No. PHR16301G
Add to Cart
Add to Cart
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Prednisolone Acetate belongs to the group of glucocorticoids, widely used to treat allergic and inflammatory conditions of the eyes.
TRUSTED_SUSTAINABILITY

Specifications

CAS 52-21-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H30O6
MDL Number MFCD00037710
Synonym Prednisolone 21-acetate; 1,4-Pregnadiene-11 beta,17 alpha,21-triol-3,20-dione 21-acetate; 11 beta,17 alpha,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate; 21-Acetoxy-1,4-pregnadiene-11 beta,17 alpha-diol-3,20-dione
Type Certified Reference Material
Quantity 1 g
Formula Weight 402.48
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.