accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNNHNNH2F

CAS RN 522629-08-9

IUPAC Name: N3-(4-fluorophenyl)-2H-pyrazolo[3,4-d]pyrimidine-3,4-diamine
Synonyms: N3-(4-fluorophenyl)-2H-pyrazolo[3,4-d]pyrimidine-3,4-diamine
Molecular Weight (g/mol): 244.23
Molecular Formula: C11H9FN6
InChi Key: UQPMANVRZYYQMD-UHFFFAOYSA-N
SMILES: NC1=NC=NC2=NNC(NC3=CC=C(F)C=C3)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results