Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CGP 57380
GREENER_CHOICESDP

Catalog No. 273150 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-315-0 50 mg
27-311 1 mg
27-311-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 27-315-0 Supplier R&D Systems™ Supplier No. 2731/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of Mnk1

Inhibitor of MAP-kinase interacting kinase-1 (Mnk1, MKNK1) (IC50 = 2.2 μM) that displays selectivity over p38, JNK1, ERK1, ERK2, PKC and c-src family kinases. Blocks phosphorylation of eIF4E in cellular assays (IC50 = 3 μM) and inhibits LPS-induced TNFα expression in macrophages.

Chemical Identifiers

CAS 522629-08-9
Molecular Formula C11H9FN6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 244.23
Purity >98%
Molecular Weight (g/mol) 244.233
Chemical Name or Material CGP 57380
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.