accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHNOCH2OF(Z)

CAS RN 524684-52-4

IUPAC Name: 4-[(2Z)-7-ethenyl-5-hydroxy-2,3-dihydro-1,3-benzoxazol-2-ylidene]-2-fluorocyclohexa-2,5-dien-1-one
Synonyms: prinaberel
Molecular Weight (g/mol): 271.25
Molecular Formula: C15H10FNO3
InChi Key: FCXYSEXZEGPLGG-DHDCSXOGSA-N
SMILES: OC1=CC2=C(O\C(N2)=C2\C=CC(=O)C(F)=C2)C(C=C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)