Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ERB 041
GREENER_CHOICESDP

Catalog No. 427650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-765-0 50 mg
42-761-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-765-0 Supplier R&D Systems™ Supplier No. 4276/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent ERβ agonist

Potent ERβ agonist; displays >200-fold selectivity for ERβ over ERα (IC50 values are 5 and 1216 nM for human ERβ and ERα; 3.1 and 620 nM for rat ERβ and ERα; and 3.7 and 750 nM for mouse ERβ and ERα respectively).

Chemical Identifiers

CAS 524684-52-4
Molecular Formula C15H10FNO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 271.24
Purity >99%
Molecular Weight (g/mol) 271.247
Chemical Name or Material ERB 041
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.