CAS RN 52485-79-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
CERILLIANT™ Buprenorphine Solution, 1.0 mg/mL in methanol (as free base), Sold by MilliporeSigma™ Supelco™
A Certified Spiking Solution™ suitable for GC/MS or LC/MS applications in pain prescription monitoring, forensic testing, or clinical toxicology.
2
CERILLIANT™ Buprenorphine Solution, 100 μg/mL in methanol, Sold by MilliporeSigma™ Supelco™
This Certified Spiking Solution™ standard is suitable for GC/MS or LC/MS applications in forensic analysis, clinical toxicology or pharmaceutical research.