CAS RN 52645-53-1
IUPAC Name:
(3-phenoxyphenyl)methyl (1R,3R)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms:
acticin ecsumin cis-(1rs)-permethrin bematin 987 dragon (1R-trans)-permethrin efmethrin permethrin, (1S-cis)- ipitox (1s-cis)-isomer permethrin
Molecular Weight (g/mol):
391.29
Molecular Formula:
C21H20Cl2O3
InChi Key:
RLLPVAHGXHCWKJ-HKUYNNGSSA-N
SMILES:
CC1(C)[C@@H](C=C(Cl)Cl)[C@H]1C(=O)OCC1=CC=CC(OC2=CC=CC=C2)=C1
1
–
1
of
1
results
1
Pesticide Mix 7, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 7 containing 8 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.