accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)CH3CH3CH3OHN(S)HOOHNHOO(R)O(S)O(E)(E)(E)(E)(E)

CAS RN 52665-74-4

IUPAC Name: (2E,4E,6R)-N-[(1S,5S,6R)-5-hydroxy-5-[(1E,3E,5E)-6-[(2-hydroxy-5-oxocyclopent-1-en-1-yl)carbamoyl]hexa-1,3,5-trien-1-yl]-2-oxo-7-oxabicyclo[4.1.0]hept-3-en-3-yl]-2,4,6-trimethyldeca-2,4-dienamide
Synonyms: manumycin
Molecular Weight (g/mol): 550.65
Molecular Formula: C31H38N2O7
InChi Key: TWWQHCKLTXDWBD-MVTGTTCWSA-N
SMILES: CCCC[C@@H](C)\C=C(/C)\C=C(/C)C(=O)NC1=C[C@@](O)(\C=C\C=C\C=C\C(=O)NC2=C(O)CCC2=O)[C@@H]2O[C@@H]2C1=O
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results