accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52698-46-1

CH3OOCH3OCH3CH3H3C

CAS RN 52698-46-1

IUPAC Name: 1,1,1-trimethoxy-2-methylpropane
Synonyms: 1,1,1-trimethoxy-2-methylpropane
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: BGLARIMANCDMQX-UHFFFAOYSA-N
SMILES: COC(OC)(OC)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results