accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3CHOOOCl

CAS RN 53-86-1

IUPAC Name: 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid
Synonyms: indomethacin methazine indo-lemmon indomethacin preparation inacid hydrochloride, indomethacin argun C19H16ClNO4 indaflex novo-methacin
Molecular Weight (g/mol): 357.79
Molecular Formula: C19H16ClNO4
InChi Key: CGIGDMFJXJATDK-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(C(=O)C3=CC=C(Cl)C=C3)C(C)=C(CC(O)=O)C2=C1
Quantity 
  • (2)
  • (1)
  • (4)
  • (5)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (5)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (5)
  • (3)
  • (9)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (5)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)