accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3HN

CAS RN 53215-95-5

IUPAC Name: benzyl[(trimethylsilyl)methyl]amine
Synonyms: benzyl[(trimethylsilyl)methyl]amine
Molecular Weight (g/mol): 193.37
Molecular Formula: C11H19NSi
InChi Key: WECLUYCAWLJMKM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CNCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)