CAS RN 5376-10-3
IUPAC Name:
(1-benzyl-1H-imidazol-2-yl)methanol
Synonyms:
(1-benzylimidazol-2-yl)methanol
Molecular Weight (g/mol):
188.23
Molecular Formula:
C11H12N2O
InChi Key:
SGIKRGDBBXWNRI-UHFFFAOYSA-N
SMILES:
OCC1=NC=CN1CC1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ Mexiletine hydrochloride
CAS: 5376-10-3 Molecular Formula: C11H12N2O