accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNHOCl

CAS RN 5437-33-2

IUPAC Name: 3,5-dichloro-1,2-dihydropyridin-2-one
Synonyms: 3,5-dichloro-1H-pyridin-2-one
Molecular Weight (g/mol): 163.99
Molecular Formula: C5H3Cl2NO
InChi Key: ZICOPWJJZSJEDL-UHFFFAOYSA-N
SMILES: ClC1=CNC(=O)C(Cl)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)