CAS RN 545-47-1
CAS RN 545-47-1
IUPAC Name:
(1R,3aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a,5a,5b,8,8,11a-hexamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
Synonyms:
lupeol
Molecular Weight (g/mol):
426.73
Molecular Formula:
C30H50O
InChi Key:
MQYXUWHLBZFQQO-JGGBMTAGSA-N
SMILES:
CC(=C)[C@@H]1CC[C@]2(C)CCC3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12
1
–
2
of
2
results
1
Thermo Scientific Chemicals Lupeol
CAS: 545-47-1 Molecular Formula: C30H50O Molecular Weight (g/mol): 426.73 MDL Number: MFCD00017351 InChI Key: MQYXUWHLBZFQQO-JGGBMTAGSA-N IUPAC Name: (1R,3aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a,5a,5b,8,8,11a-hexamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol SMILES: CC(=C)[C@@H]1CC[C@]2(C)CCC3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12