accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CONHOO(E/Z)

CAS RN 545395-94-6

IUPAC Name: 3-(4-tert-butylphenyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)prop-2-enamide
Synonyms: 3-(4-tert-butylphenyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)prop-2-enamide
Molecular Weight (g/mol): 337.42
Molecular Formula: C21H23NO3
InChi Key: GZTFUVZVLYUPRG-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=CC(=O)NC2=CC3=C(OCCO3)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)