accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OOOHN

CAS RN 546141-08-6

IUPAC Name: 3'-carbamoyl-[1,1'-biphenyl]-3-yl N-cyclohexylcarbamate
Synonyms: 3'-carbamoyl-[1,1'-biphenyl]-3-yl N-cyclohexylcarbamate
Molecular Weight (g/mol): 338.41
Molecular Formula: C20H22N2O3
InChi Key: ROFVXGGUISEHAM-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(=C1)C1=CC=CC(OC(=O)NC2CCCCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)