accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 548756-68-9

IUPAC Name: 5-{3,4-difluoro-2-[(2-fluoro-4-iodophenyl)amino]phenyl}-N-[2-(morpholin-4-yl)ethyl]-1,3,4-oxadiazol-2-amine
Synonyms: 5-{3,4-difluoro-2-[(2-fluoro-4-iodophenyl)amino]phenyl}-N-[2-(morpholin-4-yl)ethyl]-1,3,4-oxadiazol-2-amine
Molecular Weight (g/mol): 545.31
Molecular Formula: C20H19F3IN5O2
InChi Key: LZZYEMSEMRUPIM-UHFFFAOYSA-N
SMILES: FC1=C(F)C(NC2=C(F)C=C(I)C=C2)=C(C=C1)C1=NN=C(NCCN2CCOCC2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results