CAS RN 54965-24-1
Tamoxifen Citrate, USP, 99-101%, Spectrum™ Chemical
CAS: 54965-24-1 Molecular Formula: C32H37NO8 Molecular Weight (g/mol): 563.65 MDL Number: MFCD00058321 InChI Key: FQZYTYWMLGAPFJ-OQKDUQJOSA-N IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
MilliporeSigma™ Calbiochem™ Tamoxifen Citrate
Tamoxifen is a potent synthetic anti-estrogenic agent.
Thermo Scientific Chemicals Tamoxifen citrate, 98%
CAS: 54965-24-1 Molecular Formula: C32H37NO8 Molecular Weight (g/mol): 563.65 MDL Number: MFCD00058321 InChI Key: FQZYTYWMLGAPFJ-OQKDUQJOSA-N Synonym: tamoxifen citrate,istubal,zitazonium,unii-7frv7310n6,i.c.i. 46474 citrate,tamoxifen citrate salt,farmifeno,ginarsan,jenoxifen PubChem CID: 2733525 ChEBI: CHEBI:9397 IUPAC Name: 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Tamoxifen citrate
CAS: 54965-24-1 Molecular Formula: C32H37NO8 Molecular Weight (g/mol): 563.65 MDL Number: MFCD00058321 InChI Key: FQZYTYWMLGAPFJ-OQKDUQJOSA-N Synonym: tamoxifen citrate,istubal,zitazonium,unii-7frv7310n6,i.c.i. 46474 citrate,tamoxifen citrate salt,farmifeno,ginarsan,jenoxifen PubChem CID: 2733525 ChEBI: CHEBI:9397 IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Tamoxifen Citrate, USP, 99-101%, Spectrum™ Chemical
CAS: 54965-24-1 Molecular Formula: C32H37NO8 Molecular Weight (g/mol): 563.65 MDL Number: MFCD00058321 InChI Key: FQZYTYWMLGAPFJ-OQKDUQJOSA-N IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
Tamoxifen Citrate 98.0+%, TCI America™
CAS: 54965-24-1 Molecular Formula: C32H37NO8 Molecular Weight (g/mol): 563.65 MDL Number: MFCD00058321 InChI Key: FQZYTYWMLGAPFJ-OQKDUQJOSA-N Synonym: tamoxifen citrate,istubal,zitazonium,unii-7frv7310n6,i.c.i. 46474 citrate,tamoxifen citrate salt,farmifeno,ginarsan,jenoxifen PubChem CID: 2733525 ChEBI: CHEBI:9397 IUPAC Name: (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid SMILES: OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1
R&D Systems™ Tamoxifen citrate
CAS: 54965-24-1 Molecular Formula: C32H37NO8