Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tamoxifen citrate
GREENER_CHOICESDP

Catalog No. 0999100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-991-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-991-00 Supplier R&D Systems™ Supplier No. 0999/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Estrogen receptor partial agonist/antagonist

Tamoxifen citrate is an estrogen receptor antagonist/partial agonist. Selective and potent inhibitor of mammalian sterol isomerase. Neuroprotective in female rats in vivo. Also high affinity agonist at the membrane estrogen receptor GPER. Tamoxifen (Cat. No. 6432) is also available.

Chemical Identifiers

CAS 54965-24-1
Molecular Formula C32H37NO8

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 563.65
Purity >99%
Molecular Weight (g/mol) 563.65
Chemical Name or Material Tamoxifen citrate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.