accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONONNOCH3CH3HClHCl

CAS RN 55974-42-0

IUPAC Name: 2-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione dihydrochloride
Synonyms: 2-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-4,4-dimethylisoquinoline-1,3-dione dihydrochloride
Molecular Weight (g/mol): 480.43
Molecular Formula: C24H31Cl2N3O3
InChi Key: SAJKHRHHDGSJEZ-UHFFFAOYSA-N
SMILES: Cl.Cl.COC1=CC=CC=C1N1CCN(CCN2C(=O)C3=CC=CC=C3C(C)(C)C2=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results