Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ARC 239 dihydrochloride
GREENER_CHOICESDP

Catalog No. 92825 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-825 25 mg
09-285 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 92-825 Supplier R&D Systems™ Supplier No. 0928/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α2B antagonist

ARC 239 dihydrochloride is a selective α2B adrenoceptor antagonist (pKD values are 8.8, 6.7 and 6.4 at α2B, α2A, and α2D receptors respectively).

Specifications

Chemical Name or Material 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride
CAS 55974-42-0
Assay Percent Range >98%
Quantity 25 mg
Product Line Tocris
Molecular Formula C24H29N3O3.2HCl
Formula Weight Observed MW: 480.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.