accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClCl

CAS RN 56-23-5

IUPAC Name: tetrachloromethane
Synonyms: carbon tetrachloride tetrachlorkohlenstoff kohlenstofftetrachlorid tetra CCl4
Molecular Weight (g/mol): 153.81
Molecular Formula: CCl4
InChi Key: VZGDMQKNWNREIO-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)Cl
Grade 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)