CAS RN 56-54-2
phyproof™ (+)-Quinidine, ≥85% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
(+)-Quinidine
CAS: 56-54-2 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00135581,MFCD00198096 InChI Key: LOUPRKONTZGTKE-LHHVKLHASA-N Synonym: quinidine,+-quinidine,chinidin,pitayine,cin-quin,8r,9s-quinidine,conquinine,conchinin,quinidex,conchinine PubChem CID: 441074 ChEBI: CHEBI:28593 IUPAC Name: (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol SMILES: COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
Quinidine, 98%, anhydrous
CAS: 56-54-2 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00135581,MFCD00198096 InChI Key: LOUPRKONTZGTKE-LHHVKLHASA-N Synonym: quinidine,+-quinidine,chinidin,pitayine,cin-quin,8r,9s-quinidine,conquinine,conchinin,quinidex,conchinine PubChem CID: 441074 ChEBI: CHEBI:28593 SMILES: COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
Quinidine 85.0+%, TCI America™
CAS: 56-54-2 Molecular Formula: C20H24N2O2 Molecular Weight (g/mol): 324.42 MDL Number: MFCD00135581,MFCD00198096 InChI Key: LOUPRKONTZGTKE-LHHVKLHASA-N Synonym: quinidine,+-quinidine,chinidin,pitayine,cin-quin,8r,9s-quinidine,conquinine,conchinin,quinidex,conchinine PubChem CID: 441074 ChEBI: CHEBI:28593 IUPAC Name: (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol SMILES: COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
R&D Systems™ Quinidine
CAS: 56-54-2 Molecular Formula: C20H24N2O2
Sigma Aldrich Quinidine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.