Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quinidine
GREENER_CHOICESDP

Catalog No. 410850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-085-0 Supplier R&D Systems™ Supplier No. 4108/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Blocks sodium and potassium channels

Class IA antiarrythmic; reduces both Na+ and K+ channel currents, including INa, IKr and IKs. Prolongs QT and induces torsade de pointes (TdP).

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 324.42
Purity 85%
Molecular Weight (g/mol) 324.42
Chemical Name or Material Quinidine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.