CAS RN 56092-81-0
IUPAC Name:
(4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid
Synonyms:
ionomycin
Molecular Weight (g/mol):
709.02
Molecular Formula:
C41H72O9
InChi Key:
PGHMRUGBZOYCAA-OJFQOPKISA-N
SMILES:
C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Ionomycin, 96%
CAS: 56092-81-0 Molecular Formula: C41H72O9
2
R&D Systems™ Ionomycin free acid
CAS: 56092-81-0 Molecular Formula: C41H72O9