Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ionomycin free acid
GREENER_CHOICESDP

Catalog No. 20921 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-921 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 20-921 Supplier R&D Systems™ Supplier No. 2092/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Calcium ionophore

Ionomycin free acid is a calcium ionophore; more specific than A23187 .

Calcium Salt also available.

Chemical Identifiers

CAS 56092-81-0
Molecular Formula C41H72O9

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 709.01
Product Type Ionomycin Free Acid
Purity >98%
Molecular Weight (g/mol) 709.02
Chemical Name or Material Ionomycin free acid
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.