accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONFOHN

CAS RN 56177-80-1

IUPAC Name: 2-ethoxy-5-fluoro-3,4-dihydropyrimidin-4-one
Synonyms: 2-ethoxy-5-fluoro-3H-pyrimidin-4-one
Molecular Weight (g/mol): 158.13
Molecular Formula: C6H7FN2O2
InChi Key: XREDGJFKPWBNRQ-UHFFFAOYSA-N
SMILES: CCOC1=NC=C(F)C(=O)N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)