accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 563-43-9

IUPAC Name: dichloro(ethyl)alumane
Synonyms: ethylaluminium dichloride
Molecular Weight (g/mol): 126.94
Molecular Formula: C2H5AlCl2
InChi Key: UAIZDWNSWGTKFZ-UHFFFAOYSA-L
SMILES: CC[Al](Cl)Cl
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)