accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONN(E)

CAS RN 56396-35-1

IUPAC Name: (2E)-2-cyano-3-(1-phenyl-1H-indol-3-yl)prop-2-enoic acid
Synonyms: (2E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 288.31
Molecular Formula: C18H12N2O2
InChi Key: BIZNHCWFGNKBBZ-JLHYYAGUSA-N
SMILES: OC(=O)C(=C\C1=CN(C2=CC=CC=C12)C1=CC=CC=C1)\C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)