accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOFFFFF

CAS RN 565-42-4

IUPAC Name: 2,2,3,3,3-pentafluoropropyl 4-methylbenzene-1-sulfonate
Synonyms: 2,2,3,3,3-pentafluoropropyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 304.23
Molecular Formula: C10H9F5O3S
InChi Key: JBHQQXONFHOEQU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results