accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOOCH3

CAS RN 56563-37-2

IUPAC Name: but-2-yn-1-yl 4-methylbenzene-1-sulfonate
Synonyms: but-2-yn-1-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 224.27
Molecular Formula: C11H12O3S
InChi Key: HGZKJNHJABSJTC-UHFFFAOYSA-N
SMILES: CC#CCOS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results