accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NOHNBr

CAS RN 56741-95-8

IUPAC Name: 2-amino-5-bromo-6-phenyl-1,4-dihydropyrimidin-4-one
Synonyms: bropirimine
Molecular Weight (g/mol): 266.1
Molecular Formula: C10H8BrN3O
InChi Key: CIUUIPMOFZIWIZ-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C(Br)=C(N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results