accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCH3ONH3CCH3(R)OHO(S)OOHNHCH3OH3C

CAS RN 57-94-3

IUPAC Name: (1S,16R)-9,21-dihydroxy-10,25-dimethoxy-15,15,30-trimethyl-7,23-dioxa-15,30-diazaheptacyclo[22.6.2.2³,⁶.1⁸,¹².1¹⁸,²².0²⁷,³¹.0¹⁶,³⁴]hexatriaconta-3,5,8,10,12(34),18,20,22(33),24(32),25,27(31),35-dodecaene-15,30-diium dichloride
Synonyms: tubocurarine chloride 氯筒箭毒碱
Molecular Weight (g/mol): 681.65
Molecular Formula: C37H42Cl2N2O6
InChi Key: GXFZCDMWGMFGFL-KKXMJGKMSA-N
SMILES: [Cl-].[Cl-].COC1=CC2=C3[C@@H](CC4=CC=C(O)C(OC5=CC6=C(CC[NH+](C)[C@H]6CC6=CC=C(OC3=C1O)C=C6)C=C5OC)=C4)[N+](C)(C)CC2
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results