CAS RN 57432-61-8
IUPAC Name:
(2Z)-but-2-enedioic acid; (4R,7R)-N-[(2S)-1-hydroxybutan-2-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),2,9,12,14-pentaene-4-carboxamide
Synonyms:
maleic acid; methylergonovine
Molecular Weight (g/mol):
455.51
Molecular Formula:
C24H29N3O6
InChi Key:
NOFOWWRHEPHDCY-DAUURJMHSA-N
SMILES:
OC(=O)\C=C/C(O)=O.CC[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2CC3=CNC4=CC=CC(=C34)C2=C1
1
–
1
of
1
results
1
R&D Systems™ Methylergometrine maleate
CAS: 57432-61-8 Molecular Formula: C24H29N3O6