Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Methylergometrine maleate
GREENER_CHOICESDP

Catalog No. 054950 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-495-0 50 mg
05-492-50 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-495-0 Supplier R&D Systems™ Supplier No. 0549/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Active metabolite of methysergide (Cat. No. 1064)

Active metabolite of methysergide (Cat. No. 1064); uterotonic agent and oxytocic.

Chemical Identifiers

CAS 57432-61-8
Molecular Formula C24H29N3O6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 455.51
Purity >98%
Molecular Weight (g/mol) 455.511
Chemical Name or Material Methylergometrine maleate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.