accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)OHN(R)SNOONHHOOOHO

CAS RN 57564-91-7

IUPAC Name: (2S)-2-amino-4-{[(1R)-1-[(carboxymethyl)carbamoyl]-2-(nitrososulfanyl)ethyl]carbamoyl}butanoic acid
Synonyms: nitrosoglutathione s-nitroso-gsh GSNO SNOG glutathione thionitrite
Molecular Weight (g/mol): 336.32
Molecular Formula: C10H16N4O7S
InChi Key: HYHSBSXUHZOYLX-WDSKDSINSA-N
SMILES: N[C@@H](CCC(=O)N[C@@H](CSN=O)C(=O)NCC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)