CAS RN 57568-80-6
IUPAC Name:
hydrogen N-(adamantan-1-yl)-N'-cyclohexylmorpholine-4-carboximidamide chloride
Synonyms:
hydrogen N-(adamantan-1-yl)-N'-cyclohexylmorpholine-4-carboximidamide chloride
Molecular Weight (g/mol):
381.99
Molecular Formula:
C21H36ClN3O
InChi Key:
FZALCKUJYZCDOX-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].C1CCC(CC1)N=C(NC12CC3CC(CC(C3)C1)C2)N1CCOCC1
1
–
1
of
1
results
1
R&D Systems™ PNU 37883 hydrochloride
CAS: 57568-80-6 Molecular Formula: C21H36ClN3O