accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3HOH3CH3CCH3H3CCH3OH

CAS RN 57717-80-3

IUPAC Name: 2,6-di-tert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
Synonyms: 2,6-di-tert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
Molecular Weight (g/mol): 292.46
Molecular Formula: C19H32O2
InChi Key: XLWJPQQFJNGUPA-UHFFFAOYSA-N
SMILES: CC(C)(CO)CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results