accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3HNOOCH3

CAS RN 57754-86-6

IUPAC Name: hydrogen [3-(2-methoxyphenoxy)-3-phenylpropyl](methyl)amine chloride
Synonyms: hydrogen nisoxetine chloride
Molecular Weight (g/mol): 307.82
Molecular Formula: C17H22ClNO2
InChi Key: LCEURBZEQJZUPV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNCCC(OC1=CC=CC=C1OC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results