accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(R)O(S)(R)CH3(R)(S)CH3OHO(R)CH3(S)OO(R)CH3(S)HO(R)CH3O(S)CH3(S)HO(R)H3CO(S)H3CHO(R)HO(S)CH3(S)H3C(E)(E)(E)

CAS RN 579-13-5

IUPAC Name: (1S,4E,5'R,6R,6'R,7S,8R,10S,11S,12R,14S,15R,16S,18E,20E,22S,25R,27S,28R,29S)-22-ethyl-7,11,14,15-tetrahydroxy-6'-[(2S)-2-hydroxypropyl]-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
Synonyms: oligomycin A
Molecular Weight (g/mol): 791.08
Molecular Formula: C45H74O11
InChi Key: MNULEGDCPYONBU-WMBHJXFZSA-N
SMILES: CC[C@H]1CC[C@H]2O[C@@]3(CC[C@@H](C)[C@@H](C[C@H](C)O)O3)[C@H](C)[C@@H](OC(=O)\C=C\[C@@H](C)[C@H](O)[C@@H](C)C(=O)[C@@H](C)[C@H](O)[C@@H](C)C(=O)[C@@](C)(O)[C@H](O)[C@@H](C)C\C=C\C=C\1)[C@H]2C
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)