Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Oligomycin A
GSA_VAGREENER_CHOICESDP

Catalog No. 41105 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-105 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-105 Supplier R&D Systems™ Supplier No. 4110/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of mitochondrial ATPase

Oligomycin A is an inhibitor of mitochondrial ATP synthase and uncoupler of oxidative phosphorylation. Antibiotic; exhibits anti-tumor activity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 579-13-5
Molecular Formula C45H74O11

Specifications

Quantity 5 mg
Product Line Tocris
Formula Weight 791.06
Product Type Oligomycin A
Purity 98%
Molecular Weight (g/mol) 791.08
Chemical Name or Material Oligomycin A
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.