CAS RN 58-28-6
IUPAC Name:
hydrogen (3-{2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-yl}propyl)(methyl)amine chloride
Synonyms:
desipramine hydrochloride desmethylimipramine chloride desmethylimipramine monohydrochloride desimipramine, hydrochloride desipramine hcl demethylimipramine hydrochloride desmethylimipramine hydrochloride dmi hydrochloride imipraminedemethyl hydrochloride desipramine monohydrochloride
Molecular Weight (g/mol):
302.85
Molecular Formula:
C18H23ClN2
InChi Key:
XAEWZDYWZHIUCT-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CNCCCN1C2=CC=CC=C2CCC2=CC=CC=C12
1
–
1
of
1
results
1
R&D Systems™ Desipramine hydrochloride
CAS: 58-28-6 Molecular Formula: C18H23ClN2